Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EpCAM/TROP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
$200.50 - $2070.00
Specifications
| Antigen | EpCAM/TROP1 |
|---|---|
| Clone | 158206 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Flow Cytometry, CyTOF, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9601SP
![]() |
R&D Systems
MAB9601SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9601100
![]() |
R&D Systems
MAB9601100 |
100 μg |
Each for $573.00
|
|
|||||
MAB9601500
![]() |
R&D Systems
MAB9601500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| EpCAM/TROP1 | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4072 | |
| Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1 Extracellular domain | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human EpCAM/TROP-1 in ELISAs. |
| 158206 | |
| Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| EPCAM | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title