Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EpCAM/TROP-1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB960P100
This item is not returnable.
View return policy
Description
EpCAM/TROP1 Polyclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry.Specifications
EpCAM/TROP1 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CAA32870 | |
Goat | |
Affinity Purified | |
RUO | |
4072 | |
Human | |
IgG |
Flow Cytometry | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1, extracellular domain Gln24-Lys265 Accession # CAA32870 | |
100 Tests | |
Primary | |
Detects human EpCAM/TROP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhMCAM, rhNCAM L1, and recombinant mouse OCAM is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction