Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EpCAM/TROP1 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9601U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9601U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9601U100 Supplier R&D Systems Supplier No. FAB9601U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EpCAM/TROP1
Applications Flow Cytometry
Classification Monoclonal
Clone 158206
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Gene Symbols EPCAM
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1 Extracellular domain
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4072
Test Specificity Detects human EpCAM/TROP-1 in ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.