Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EphA1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB638
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB638 500 μg
MAB638SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB638 Supplier R&D Systems Supplier No. MAB638
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EphA1 Monoclonal specifically detects EphA1 in Human samples. It is validated for Western Blot.

Specifications

Antigen EphA1
Applications Western Blot
Classification Monoclonal
Clone 86810
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P21709
Gene Alias EC 2.7.10, EC 2.7.10.1, EPH receptor A1, eph tyrosine kinase 1, EphA1, EPHephrin type-A receptor 1, EPHT, EPHT1erythropoietin-producing hepatoma amplified sequence, Esk, MGC163163, oncogene EPH, soluble EPHA1 variant 1, soluble EPHA1 variant 2, Tyrosine-protein kinase receptor EPH
Gene Symbols EPHA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphA1 Arg24-Glu547 (predicted) Accession # P21709
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2041
Test Specificity Detects human EphA1 in direct ELISAs and Western blots. No cross-reactivity with recombinant mouse EphA2, A3, A4, A6, A7, A8, B2, B4, B5, B6, recombinant rat (rr) EphA5, rrEphB1, or recombinant human EphB1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.