Learn More
Human EphA2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | EphA2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 371805 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 350 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P29317 |
| Gene Alias | ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.