Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EphA2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3035R100UG
This item is not returnable.
View return policy
Description
EphA2 Monoclonal specifically detects EphA2 in Human samples. It is validated for Flow Cytometry.Specifications
EphA2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P29317 | |
EPHA2 | |
Mouse myeloma cell line NS0-derived recombinant human EphA2 Gln25-Asn534 Accession # P29317 | |
100 μg | |
Primary | |
Detects human EphA2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse EphA4, A5, A6, A7, A8, or recombinant rat EphB1 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
371805 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1969 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction