Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EphA2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3035
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3035 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3035 Supplier R&D Systems Supplier No. BAF3035
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

EphA2 Polyclonal specifically detects EphA2 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen EphA2
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells
Gene Accession No. P29317
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Gene Symbols EPHA2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphA2 Gln25-Asn534 Accession # P29317
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1969
Test Specificity Detects human EphA2 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse (rm) EphA2 is observed and less than 1% cross-reactivity with recombinant human (rh) EphA1, rmEphA4, rhEphA5, rmEphA6, rmEphA7, and rmEphA8 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.