Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EphB3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5667SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5667SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5667SP Supplier R&D Systems Supplier No. MAB5667SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EphB3 Monoclonal specifically detects EphB3 in Human samples. It is validated for Western Blot.

Specifications

Antigen EphB3
Applications Western Blot
Classification Monoclonal
Clone 647308
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P54753
Gene Alias Cek10, EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Mdk5, Sek4, Tyro6, Tyrosine-protein kinase TYRO6
Gene Symbols EPHB3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphB3 Leu38-Ala550 Accession # P54753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2049
Test Specificity Detects human EphB3 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant mouse (rm) EphB3, 25% cross-reactivity with rmEphB2, and no cross-reactivity with recombinant human (rh) EphA1, A2, A5, A6, A10, rmEphA3, A4, A7, B4, B6, or recombinant rat EphB1.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.