Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EphB3 Antibody, R&D Systems™

Catalog No. AF5667
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5667 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5667 Supplier R&D Systems Supplier No. AF5667
This item is not returnable. View return policy

Sheep Polyclonal Antibody

EphB3 Polyclonal specifically detects EphB3 in Human samples. It is validated for Western Blot.

Specifications

Antigen EphB3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P54753
Gene Alias Cek10, EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Mdk5, Sek4, Tyro6, Tyrosine-protein kinase TYRO6
Gene Symbols EPHB3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphB3 Leu38-Ala550 Accession # P54753
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2049
Test Specificity Detects human EphB3 in direct ELISAs and Western blots. In direct ELISAs, approximately 3% cross-reactivity with recombinant mouse EphB3 is observed, and less than 1% cross-reactivity with recombinant rat EphB1, recombinant human (rh) EphB2 and rhEphB4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.