Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EphB4 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB3038U100UG
This item is not returnable.
View return policy
Description
EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for Flow Cytometry.Specifications
EphB4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAH52804 | |
EPHB4 | |
Mouse myeloma cell line NS0-derived recombinant human EphB4 Ala16-Ala539 Accession # AAH52804 | |
RUO | |
2050 | |
Human | |
IgG1 |
Flow Cytometry | |
395810 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, Htk, HTKephrin receptor EphB4, Mdk2, Myk1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
Rat | |
100 μg | |
Primary | |
Detects human EphB4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human EphA1, A2, A5, A6, A10, B2, B3, B6, recombinant mouse EphA3, A4, or recombinant rat EphB1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction