Learn More
Human EphB4 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 7 publications
Supplier:  R&D Systems AF3038
Catalog No. AF3038
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
EphB4 Polyclonal specifically detects EphB4 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, CyTOF-ready, Knockout Validated.
Specifications
Specifications
EphB4
Polyclonal
Unconjugated
P54760
EPHB4
Mouse myeloma cell line NS0-derived recombinant human EphB4 Leu16-Ala539 Accession # P54760
100 μg
Primary
Detects human EphB4 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse (rm) EphB4 is observed and less than 1% cross-reactivity with recombinant human (rh) EphA4, rhEphA3, rhEphB2, and rhEphB3 is observed.
Human
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
LYOPH
Western Blot 2 ug/mL, Simple Western 50 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, Htk, HTKephrin receptor EphB4, Mdk2, Myk1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11
Goat
Affinity Purified
RUO
2050
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.