Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EphB6 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA33841T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA33841T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA33841T100 Supplier R&D Systems Supplier No. FAB33841T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EphB6
Applications Flow Cytometry
Classification Monoclonal
Clone 465327
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O15197
Gene Alias EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911
Gene Symbols EPHB6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphB6 Val17-Thr579 Accession # O15197
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2051
Test Specificity Detects human EphB6 in direct ELISAs.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.