Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EphB6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB33841SP
This item is not returnable.
View return policy
Description
EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
EphB6 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
O15197 | |
EPHB6 | |
Mouse myeloma cell line NS0-derived recombinant human EphB6 Val17-Thr579 Accession # O15197 | |
25 μg | |
Primary | |
Detects human EphB6 in direct ELISAs. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
465327 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, Hep, HEPTyrosine-protein kinase-defective receptor EPH-6, Mep, MGC129910, MGC129911 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2051 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction