Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Ephrin-B2 ELISA Kit (Colorimetric)
SDP

Catalog No. NB171836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171836 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171836 Supplier Novus Biologicals™ Supplier No. NBP318214
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.3125-20 ng/ml
Assay Sensitivity 0.14 ng/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Cell Lysates, Tissue Homogenates
For Use With (Application) ELISA
Gene ID (Entrez) 1948
Gene Symbol EFNB2
Quantity 1 Kit
Research Discipline Neuroscience, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ephrin B2, EPLG5, Htk-L, HTKL, LERK5
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.