Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ephrin-B3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF395
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF395 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF395 Supplier R&D Systems Supplier No. AF395
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Ephrin-B3 Polyclonal specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Ephrin-B3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q15768
Gene Alias EFL-6, Efnb3, Elk-L3, EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EphrinB3, Epl8, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6, NLERK-2
Gene Symbols EFNB3
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Ephrin-B3 Leu28-Ser224 Accession # Q15768
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1949
Test Specificity Detects human Ephrin-B3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Ephrin-A5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.