Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ephrin-B3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF395
This item is not returnable.
View return policy
Description
Ephrin-B3 Polyclonal specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot.Specifications
Ephrin-B3 | |
Polyclonal | |
Biotin | |
Q15768 | |
EFNB3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Ephrin-B3 Leu28-Ser224 Accession # Q15768 | |
50 μg | |
Primary | |
Detects human Ephrin-B3 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-A1, rmEphrin-A2, recombinant human (rh) Ephrin-A3, rhEphrin-A4, rhEphrin-A5, rmEphrin-B1, and rmEphrin-B2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
EFL-6, Efnb3, Elk-L3, EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EphrinB3, Epl8, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6, NLERK-2 | |
Goat | |
Affinity Purified | |
RUO | |
1949 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction