Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Epiregulin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC14251V100UG
This item is not returnable.
View return policy
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Epiregulin | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
O14944 | |
EREG | |
E. coli-derived recombinant human Epiregulin Val63-Leu108 Accession # O14944 | |
RUO | |
2069 | |
Human | |
IgG2b |
Flow Cytometry | |
183629 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
epiregulin, ER, EREG, proepiregulin | |
Mouse | |
100 μg | |
Primary | |
Detects human Epiregulin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Betacellulin, rhEGF, recombinant mouse Epiregulin, or rhHB-EGF is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction