Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Epiregulin Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC14251N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC14251N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC14251N100 Supplier R&D Systems Supplier No. IC14251N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Epiregulin
Applications Flow Cytometry
Classification Monoclonal
Clone 183629
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O14944
Gene Alias epiregulin, ER, EREG, proepiregulin
Gene Symbols EREG
Host Species Mouse
Immunogen E. coli-derived recombinant human Epiregulin Val63-Leu108 Accession # O14944
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2069
Test Specificity Detects human Epiregulin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Betacellulin, rhEGF, recombinant mouse Epiregulin, or rhHB-EGF is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.