Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Epiregulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $1929.00
Specifications
| Antigen | Epiregulin |
|---|---|
| Clone | 183625 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1425SP
![]() |
R&D Systems
MAB1425SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1425100
![]() |
R&D Systems
MAB1425100 |
100 μg |
Each for $526.00
|
|
|||||
MAB1425500
![]() |
R&D Systems
MAB1425500 |
500 μg |
Each for $1,929.00
|
|
|||||
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Western Blot.Specifications
| Epiregulin | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| epiregulin, ER, EREG, proepiregulin | |
| EREG | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 183625 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| O14944 | |
| 2069 | |
| E. coli-derived recombinant human Epiregulin Val63-Leu108 Accession # O14944 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects the EGF-like domain of human Epiregulin in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 100% cross-reactivity with recombinant mouse Epiregulin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title