Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Epiregulin Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | Epiregulin |
---|---|
Clone | 870126 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB14252SP
![]() |
R&D Systems
MAB14252SP |
25 μg |
Each for $200.50
|
|
|||||
MAB14252100
![]() |
R&D Systems
MAB14252100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for ELISA.Specifications
Epiregulin | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
O14944 | |
2069.0 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
870126 | |
ELISA | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
epiregulin, ER, EREG, proepiregulin | |
E. coli-derived recombinant human Epiregulin, Val63-Leu108, Accession No. O14944 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Epiregulin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title