Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Eps15 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC8480T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8480T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC8480T100 Supplier R&D Systems Supplier No. IC8480T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EPS15 Monoclonal specifically detects EPS15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen EPS15
Applications Flow Cytometry
Classification Monoclonal
Clone 1261C
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P42566
Gene Alias AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15
Host Species Rabbit
Immunogen E.coli-derived recombinant human Eps15, Ala448-Thr579, Accession # P42566
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2060
Test Specificity Detects human Eps15 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.