Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Eps15 Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF8480SP
This item is not returnable.
View return policy
Description
EPS15 Polyclonal specifically detects EPS15 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
EPS15 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
2060 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.05 μg/mL, Simple Western 2.5 μg/mL, Immunocytochemistry 5-15 μg/mL | |
P42566 | |
EPS15 | |
E. coli-derived recombinant human Eps15 Met492-Thr579 Accession # P42566 | |
25 μg | |
Primary | |
Detects human Eps15 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction