Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPS8L2 Full-length ORF (NP_073609.2, 1 a.a. - 715 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89943495
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the EPS8 gene family. The encoded protein, like other members of the family, is thought to link growth factor stimulation to actin organization, generating functional redundancy in the pathways that regulate actin cytoskeletal remodeling. [provided by RefSeq]

Sequence: MSQSGAVSCCPGATNGSLGRSDGVAKMSPKDLFEQRKKYSNSNVIMHETSQYHVQHLATFIMDKSEAITSVDDAIRKLVQLSSKEKIWTQEMLLQVNDQSLRLLDIESQEELEDFPLPTVQRSQTVLNQLRYPSVLLLVCQDSEQSKPDVHFFHCDEVEAELVHEDIESALADCRLGKKMRPQTLKGHQEKIRQRQSILPPPQGPAPIPFQHRGGDSPEAKNRVGPQVPLSEPGFRRRESQEEPRAVLAQKIEKETQILNCALDDIEWFVARLQKAAEAFKQLNQRKKGKKKGKKAPAEGVLTLRARPPSEGEFIDCFQKIKLAINLLAKLQKHIQNPSAAELVHFLFGPLDLIVNTCSGPDIARSVSCPLLSRDAVDFLRGHLVPKEMSLWESLGESWMRPRSEWPREPQVPLYVPKFHSGWEPPVDVLQEAPWEVEGLASAPIEEVSPVSRQSIRNSQKHSPTSEPTPPGDALPPVSSPHTHRGYQPTPAMAKYVKILYDFTARNANELSVLKDEVLEVLEDGRQWWKLRSRSGQAGYVPCNILGEARPEDAGAPFEQAGQKYWGPASPTHKLPPSFPGNKDELMQHMDEVNDELIRKISNIRAQPQRHFRVERSQPVSQPLTYESGPDEVRAWLEAKAFSPRIVENLGILTGPQLFSLNKEELKKVCGEEGVRVYSQLTMQKAFLEKQQSGSELEELMNKFHSMNQRRGEDS

Specifications

Accession Number NP_073609.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 64787
Molecular Weight (g/mol) 107kDa
Name EPS8L2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MSQSGAVSCCPGATNGSLGRSDGVAKMSPKDLFEQRKKYSNSNVIMHETSQYHVQHLATFIMDKSEAITSVDDAIRKLVQLSSKEKIWTQEMLLQVNDQSLRLLDIESQEELEDFPLPTVQRSQTVLNQLRYPSVLLLVCQDSEQSKPDVHFFHCDEVEAELVHEDIESALADCRLGKKMRPQTLKGHQEKIRQRQSILPPPQGPAPIPFQHRGGDSPEAKNRVGPQVPLSEPGFRRRESQEEPRAVLAQKIEKETQILNCALDDIEWFVARLQKAAEAFKQLNQRKKGKKKGKKAPAEGVLTLRARPPSEGEFIDCFQKIKLAINLLAKLQKHIQNPSAAELVHFLFGPLDLIVNTCSGPDIARSVSCPLLSRDAVDFLRGHLVPKEMSLWESLGESWMRPRSEWPREPQVPLYVPKFHSGWEPPVDVLQEAPWEVEGLASAPIEEVSPVSRQSIRNSQKHSPTSEPTPPGDALPPVSSPHTHRGYQPTPAMAKYVKILYDFTARNANELSVLKDEVLEVLEDGRQWWKLRSRSGQAGYVPCNILGEARPEDAGAPFEQAGQKYWGPASPTHKLPPSFPGNKDELMQHMDEVNDELIRKISNIRAQPQRHFRVERSQPVSQPLTYESGPDEVRAWLEAKAFSPRIVENLGILTGPQLFSLNKEELKKVCGEEGVRVYSQLTMQKAFLEKQQSGSELEELMNKFHSMNQRRGEDS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias EPS8R2/FLJ16738/FLJ21935/FLJ22171/MGC126530/MGC3088
Common Name EPS8L2
Gene Symbol EPS8L2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.