Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Equine 5'-Nucleotidase/CD73 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5795T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5795T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5795T100 Supplier R&D Systems Supplier No. FAB5795T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human, Equine samples. It is validated for Flow Cytometry.

Specifications

Antigen 5'-Nucleotidase/CD73
Applications Flow Cytometry
Classification Monoclonal
Clone 606112
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P21589
Gene Alias 5' nucleotidase (CD73), 5'-NT, 5-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NT5E, NTE, Purine 5-Prime-Nucleotidase
Gene Symbols NT5E
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human 5'-Nucleotidase/CD73 Trp27-Ile511 Accession # P21589
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4907
Test Specificity Detects recombinant human 5'-Nucleotidase in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse 5'-Nucleotidase is observed.
Target Species Human, Equine
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.