Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Equine 5'-Nucleotidase/CD73 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5795R100UG
This item is not returnable.
View return policy
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human, Equine samples. It is validated for Flow Cytometry.Specifications
5'-Nucleotidase/CD73 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P21589 | |
NT5E | |
Chinese hamster ovary cell line CHO-derived recombinant human 5'-Nucleotidase/CD73 Trp27-Ile511 Accession # P21589 | |
RUO | |
4907 | |
Human, Equine | |
IgG2b |
Flow Cytometry | |
606112 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
5' nucleotidase (CD73), 5'-NT, 5-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NT5E, NTE, Purine 5-Prime-Nucleotidase | |
Mouse | |
100 μg | |
Primary | |
Detects recombinant human 5'-Nucleotidase in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse 5'-Nucleotidase is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction