Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ER alpha/NR3A1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $491.00
Specifications
Antigen | ER alpha/NR3A1 |
---|---|
Clone | 2424B |
Dilution | Immunohistochemistry 10-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB57153SP
![]() |
R&D Systems
MAB57153SP |
25 μg |
Each for $214.00
|
|
|||||
MAB57153100
![]() |
R&D Systems
MAB57153100 |
100 μg |
Each for $491.00
|
|
|||||
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
ER alpha/NR3A1 | |
Immunohistochemistry 10-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P03372 | |
2099.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2424B | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ER, ER alpha, Era, ER-alpha, ESR1, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
E. coli-derived recombinant human ER alpha/NR3A1, Met1-Gln116, Accession No. P03372 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ER alpha/NR3A1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title