Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ER alpha/NR3A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems PPH462400
This item is not returnable.
View return policy
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Direct ELISA.Specifications
ER alpha/NR3A1 | |
Monoclonal | |
Unconjugated | |
A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
ESR1 | |
Recombinant human ER alpha/NR3A1 aa 2-180 | |
100 μg | |
Primary | |
This antibody specifically recognizes human ER alpha and cross-reacts with mouse and rat ER alpha . This antibody does not recognize human ER beta . Not yet tested in other species. | |
This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA | |
H4624 | |
Western Blot, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation, Direct ELISA | |
ER, ER alpha, Era, ER-alpha, ESR1, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2099 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction