Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ER beta/NR3A2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC7106N100UG
This item is not returnable.
View return policy
Description
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
ER beta/NR3A2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q92731 | |
ESR2 | |
E. coli-derived recombinant human ER beta/NR3A2 Met1-Gly318 Accession # Q92731 | |
RUO | |
2100 | |
Human | |
IgG1 |
Flow Cytometry | |
733930 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Erb, ER-beta, ESR2, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
Mouse | |
100 μg | |
Primary | |
Detects human ER beta/NR3A2 in direct ELISAs and Western blot. In direct ELISA and Western blot, no cross-reactivity with recombinant human ER alpha is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction