Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ErbB2/Her2 (Based on Research Grade Trastuzumab Biosimilar) Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $538.00
Specifications
Antigen | ErbB2/Her2 |
---|---|
Clone | H2M5B |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Inhibition of Cell Growth 20-100 ng/mL, CyTOF-ready |
Applications | Flow Cytometry, Inhibition Assays, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9896SP
![]() |
R&D Systems
MAB9896SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9896100
![]() |
R&D Systems
MAB9896100 |
100 μg |
Each for $538.00
|
|
|||||
Description
ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, Inhibition of Cell Growth, CyTOF-ready.Specifications
ErbB2/Her2 | |
Flow Cytometry 0.25 ug/10^6 cells, Inhibition of Cell Growth 20-100 ng/mL, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, Neu Oncogene, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGL, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, TKR1, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
Human ErbB2/Her2 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human ErbB2/Her2 in Western blots. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibodyTrastuzumab and uses a mouse IgG1 backbone structure. This product is for research use only. |
H2M5B | |
Flow Cytometry, Inhibition Assays, CyTOF | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
2064 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title