Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ErbB3/Her3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
$200.50 - $2020.00
Specifications
Antigen | ErbB3/Her3 |
---|---|
Clone | 66223 |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.0075-0.03 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Flow Cytometry, Neutralization, CyTOF, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3481SP
![]() |
R&D Systems
MAB3481SP |
25 μg |
Each for $200.50
|
|
|||||
MAB3481100
![]() |
R&D Systems
MAB3481100 |
100 μg |
Each for $573.00
|
|
|||||
MAB3481500
![]() |
R&D Systems
MAB3481500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
ErbB3/Her3 | |
Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.0075-0.03 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P21860 | |
2065 | |
Mouse myeloma cell line NS0-derived recombinant human ErbB3/Her3 Ser20-Thr643 Accession # P21860 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ErbB3/Her3 in ELISAs. Does not cross-react with recombinant EGF R. |
66223 | |
Flow Cytometry, Neutralization, CyTOF, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
ERBB3 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title