Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ErbB4/Her4 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB11311N100
This item is not returnable.
View return policy
Description
ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human samples. It is validated for Flow Cytometry.Specifications
ErbB4/Her4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 5 uL/10^6 cells | |
Q15303 | |
ERBB4 | |
Mouse myeloma cell line NS0-derived recombinant human ErbB4/Her4 Glu26-Arg649 Accession # Q15303 | |
100 Tests | |
Primary | |
Detects human ErbB4/Her4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) EGF R, rhErbB2, or rhErbB3 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
182818 | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2066 | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction