Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ErbB4/Her4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11311SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11311SP 25 μg
MAB11311 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11311SP Supplier R&D Systems Supplier No. MAB11311SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen ErbB4/Her4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 182818
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q15303
Gene Alias avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian)
Gene Symbols ERBB4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ErbB4/Her4 Glu26-Arg649 Accession # Q15303
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2066
Test Specificity Detects human ErbB4/Her4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) EGF R, rhErbB2, or rhErbB3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.