Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ERK1 Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF1940
This item is not returnable.
View return policy
Description
ERK1 Polyclonal specifically detects ERK1 in Human samples. It is validated for Western Blot.Specifications
ERK1 | |
Polyclonal | |
Unconjugated | |
P27361 | |
MAPK3 | |
Human ERK1 synthetic peptide Met1-Val28 Accession # P27361 | |
50 μg | |
Primary | |
Detects human ERK1 in Western blots. Because the peptide immunogen corresponds to a region of ERK1 largely absent in ERK2, no reactivity with ERK2 is present. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MAPK3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, P44MAPK, PRKM3, PRKM3EC 2.7.11.24 | |
Rabbit | |
Affinity Purified | |
RUO | |
5595 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction