Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ERMAP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4928SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4928SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4928SP Supplier R&D Systems Supplier No. MAB4928SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ERMAP Monoclonal specifically detects ERMAP in Human samples. It is validated for Western Blot.

Specifications

Antigen ERMAP
Applications Western Blot
Classification Monoclonal
Clone 767504
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96PL5
Gene Alias erythroblast membrane-associated protein, erythroblast membrane-associated protein (RD and SC blood groups), erythroblast membrane-associated protein (Scianna blood group), erythroid membrane-associated protein, hERMAP, MGC118810, MGC118811, PRO2801, Radin blood group, Radin blood group (Rd), Radin blood group antigen, RD, RDMGC118812, SC, Scianna blood group, Scianna blood group (Sc), Scianna blood group antigen, SCMGC118813
Gene Symbols ERMAP
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ERMAP His30-Ala155 Accession # Q96PL5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114625
Test Specificity Detects human ERMAP in direct ELISAS.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.