Learn More
Human ERR alpha 1/NR3B1 Antibody (Clone H5844), R&D Systems™

Description
Specifications
Specifications
| Antigen | ERR alpha/NR3B1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Clone | H5844 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation, Direct ELISA |
| Formulation | A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide |
| Gene Alias | ERR1, ERR1Estrogen receptor-like 1, ERRa, ESRL1, ESRL1ERR-alpha, ESRRA, estrogen-related receptor alphaERRalpha, NR3B1, NR3B1Nuclear receptor subfamily 3 group B member 1, steroid hormone receptor ERR1 |
| Gene Symbols | ESRRA |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.