Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™

Mouse Monoclonal Antibody
$591.44 - $2171.79
Specifications
| Antigen | Erythropoietin/EPO |
|---|---|
| Clone | AE7A5 |
| Dilution | Western Blot 1 ug/mL, Immunoaffinity Purification |
| Applications | Western Blot, Affinity Purification |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2871100
![]() |
R&D Systems
MAB2871100 |
100 μg |
Each for $591.44
|
|
|||||
MAB2871500
![]() |
R&D Systems
MAB2871500 |
500 μg |
Each for $2,171.79
|
|
|||||
|
MAB2871SP
|
R&D Systems
MAB2871SP |
25 μg | N/A | N/A | N/A | ||||
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, Immunoaffinity Purification.Specifications
| Erythropoietin/EPO | |
| Western Blot 1 ug/mL, Immunoaffinity Purification | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01588 | |
| 2056 | |
| Erythropoietin/EPO synthetic peptide analog, Ala28-Thr53, Accession # P01588 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. |
| AE7A5 | |
| Western Blot, Affinity Purification | |
| Unconjugated | |
| RUO | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| EPO | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Recognizes an epitope within the first 26 amino acids at the NH2 terminus of human urinary and human recombinant EPO. Binds human and rat EPO but does not bind sheep, baboon, or dog EPO (1, 2). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title