Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $351.50
Specifications
Antigen | Erythropoietin/EPO |
---|---|
Clone | 971019 |
Dilution | Neutralization 0.02-0.2 μg;/mL |
Applications | Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2872SP
![]() |
R&D Systems
MAB2872SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2872100
![]() |
R&D Systems
MAB2872100 |
100 μg |
Each for $351.50
|
|
|||||
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Neutralization.Specifications
Erythropoietin/EPO | |
Neutralization 0.02-0.2 μg;/mL | |
Monoclonal | |
Supernatant | |
Human | |
CAA26094 | |
2056.0 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
971019 | |
Neutralization | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Chinese Hamster Ovary cell line CHO-derived human Erythropoietin/EPO protein, Ala28-Arg193, Accession # CAA26094 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Erythropoietin/EPO in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title