Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Erythropoietin Quantikine IVD ELISA Kit

Catalog No. dep00
Click to view available options
Sufficient For:
1 x 96-well Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Human Erythropoietin Quantikine IVD ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Erythropoietin Quantikine IVD ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Erythropoietin in serum (100 ul), edta plasma (100 ul).

Specifications

Assay Range 2.50 - 200 mIU/mL
Assay Sensitivity 0.6 mIU/mL
Conjugate HRP
Sample Type Serum
Specificity Natural and Recombinant Human Epo
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 2056
Sample Volume Serum (100uL),EDTA Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Target Erythropoietin/EPO
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.