Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Erythropoietin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $1842.00
Specifications
| Antigen | Erythropoietin R |
|---|---|
| Clone | 38407 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3071SP
![]() |
R&D Systems
MAB3071SP |
25 μg |
Each for $200.50
|
|
|||||
MAB3071100
![]() |
R&D Systems
MAB3071100 |
100 μg |
Each for $502.50
|
|
|||||
MAB3071500
![]() |
R&D Systems
MAB3071500 |
500 μg |
Each for $1,842.00
|
|
|||||
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Erythropoietin R | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P19235 | |
| 2057 | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Erythropoietin Receptor (Epo R) in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Epo, recombinant mouse Epo R, or rhThrombopoietin R was observed. |
| 38407 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| EPOR | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title