Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Erythropoietin R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB307500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB307500 500 μg
MAB307SP 25 μg
MAB307100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB307500 Supplier R&D Systems Supplier No. MAB307500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.

Specifications

Antigen Erythropoietin R
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 38409
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P19235
Gene Alias EpoR, EPO-R, erythropoietin receptor, MGC138358
Gene Symbols EPOR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2057
Test Specificity Detects human Erythropoietin R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Erythropoietin R is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.