Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Erythropoietin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Erythropoietin R |
---|---|
Clone | 1052708 |
Dilution | Immunocytochemistry 3-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11190SP
![]() |
R&D Systems
MAB11190SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11190100
![]() |
R&D Systems
MAB11190100 |
100 μg |
Each for $468.00
|
|
|||||
Description
Erythropoietin R Monoclonal antibody specifically detects Erythropoietin R in Human samples. It is validated for ImmunocytochemistrySpecifications
Erythropoietin R | |
Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R, Ala25-Pro250, Accession # P19235 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1052708 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P19235 | |
2057 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title