Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Erythropoietin R Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB307F
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Flow Cytometry.Specifications
Erythropoietin R | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
2057 | |
Human | |
Purified |
Flow Cytometry | |
38409 | |
Flow Cytometry 10 uL/10^6 cells | |
P19235 | |
EPOR | |
Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235 | |
100 Tests | |
Primary | |
Detects human Erythropoietin R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Erythropoietin R is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction