Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ETV1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
Antigen | ETV1 |
---|---|
Clone | 815207 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9389SP
![]() |
R&D Systems
MAB9389SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9389100
![]() |
R&D Systems
MAB9389100 |
100 μg |
Each for $549.50
|
|
|||||
Description
ETV1 Monoclonal specifically detects ETV1 in Human samples. It is validated for Western Blot.Specifications
ETV1 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P50549 | |
2115 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
815207 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
DKFZp781L0674, ER81, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534 | |
E. coli-derived recombinant human ETV1 Asn120-Leu190 Accession # P50549 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ETV1 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title