Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ETV5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7107SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7107SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7107SP Supplier R&D Systems Supplier No. MAB7107SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ETV5/ERM Monoclonal specifically detects ETV5/ERM in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen ETV5/ERM
Applications Immunocytochemistry
Classification Monoclonal
Clone 654404
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41161
Gene Alias ERM, ERMEts-related protein ERM, ETS translocation variant 5, ets variant 5, ets variant gene 5 (ets-related molecule), ets-related molecule
Gene Symbols ETV5
Host Species Mouse
Immunogen E. coli-derived recombinant human ETV5 Asn119-Ser223 Accession # P41161
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2119
Test Specificity Detects human ETV5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) ETV1 or rhETV2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.