Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human EVI-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB75061SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB75061SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB75061SP Supplier R&D Systems Supplier No. MAB75061SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EVI-1 Monoclonal specifically detects EVI-1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen EVI-1
Applications Immunocytochemistry
Classification Monoclonal
Clone 702321
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q03112
Gene Alias AML1-EVI-1, AML1-EVI-1 fusion protein, ecotropic viral integration site 1, Ecotropic virus integration site 1 protein homolog, EVI1, EVI-1, EVI1MDS1-EVI1, MDS1, MDS1 and EVI1 complex locus, MDS1 and EVI1 complex locus protein EVI1, MDS1 and EVI1 complex locus protein MDS1, MECOM, MGC163392, MGC97004, myelodysplasia syndrome 1, Myelodysplasia syndrome 1 protein, Myelodysplasia syndrome-associated protein 1, oncogene EVI1, PRDM3, zinc finger protein Evi1
Gene Symbols MECOM
Host Species Mouse
Immunogen E. coli-derived recombinant human EVI-1 Gly241-Met430 Accession # Q03112
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2122
Test Specificity Detects human EVI-1 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.