Learn More
Novus Biologicals™ Human Exosome (Cell Media) ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Sensitivity | <0.35 μg of overall exosome or < 50 pg targeted exosomes protein |
| Immunoassay Kit Format | Colorimetric Detection Kit |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA, Immunoassay |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. After reconstitution store at -70°C. |
| Synonym | EV detection, exosome detection, extracellular vesicle detection |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.