Learn More
Specifications
Specifications
| Assay Sensitivity | < 0.35 ug of overall exosome or < 50 pg targeted exosomes protein |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Urine |
| For Use With (Application) | ELISA, Immunoassay |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4C. |
| Synonym | EV detection, exosome detection, extracellular vesicle detection |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
