Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Exostosin-like 3/EXTL3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Exostosin-like 3/EXTL3 |
---|---|
Clone | 311007 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Exostosin-like 3/EXTL3 Monoclonal specifically detects Exostosin-like 3/EXTL3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Exostosin-like 3/EXTL3 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
O43909 | |
2137 | |
Mouse myeloma cell line NS0-derived recombinant human Exostosin-like 3/EXTL3 Thr52-Ile919 Accession # O43909 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Exostosin-like 3/EXTL3 in direct ELISAs and Western blots. |
311007 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
botv, DKFZp686C2342, EC 2.4.1.223, exostoses (multiple)-like 3, Exostosin like 3, exostosin-like 3, EXTL1L, EXTL3, EXTR1BOTV, EXT-related protein 1, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Hereditary multiple exostoses gene isolog, KIAA0519, Multiple exostosis-like protein 3, Putative tumor suppressor protein EXTL3, REG, reg receptor, REGR, RPR | |
EXTL3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title