Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EZFIT aa 6-199 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems PPK971600
This item is not returnable.
View return policy
Description
EZFIT/ZNF71 Monoclonal specifically detects EZFIT/ZNF71 in Human samples. It is validated for Western Blot, Direct ELISA.Specifications
EZFIT/ZNF71 | |
Monoclonal | |
Unconjugated | |
A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
ZNF71 | |
aa 6-199 | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
K9716 | |
Western Blot, Direct ELISA | |
Cos26, endothelial zinc finger protein induced by tumor necrosis factor alpha, Kruppel-related zinc finger protein, zinc finger protein 71 (Cos26), zinc finger protein 71EZFITCos26, ZNF47, ZNF71 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
58491 | |
Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction