Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human F-box protein 15/FBXO15 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC6035R100UG
This item is not returnable.
View return policy
Description
F-box protein 15/FBXO15 Monoclonal specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
F-box protein 15/FBXO15 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8NCQ5-2 | |
FBXO15 | |
E. coli-derived recombinant human F-box protein 15/FBXO15 isoform 2 Asn298-Tyr434 Accession # Q8NCQ5-2 | |
RUO | |
201456 | |
Human | |
IgG2a |
Flow Cytometry | |
612834 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
F-box only protein 15, Fbox protein 15, F-box protein 15, FBX15, FBX15MGC39671, FBXO15 | |
Mouse | |
100 μg | |
Primary | |
Detects human F-box protein 15/FBXO15 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse F-box protein 15 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction